Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mex-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306334.3 | NP_001293263.1 | 1 - 2 | 289 | 70 | |
NM_001381229.1 | NP_001367884.1 | 6 - 7 | 1098 | 70 | ||
NM_058280.9 | NP_490681.2 | 6 - 7 | 1090 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |