Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bli-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058285.5 | NP_490686.3 | 14 - 15 | 3250 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |