Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mbtr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129070.2 | NP_001122542.1 | 1 - 2 | 121 | 93 | |
GenBank mRNA | DQ904352.1 | - | 1 - 2 | 61 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |