Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnp-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383945.1 | 6 - 7 | 1463 | 60 | ||
NM_058323.5 | NP_490724.1 | 5 - 6 | 1563 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |