Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hum-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261950.4 | NP_001248879.1 | 4 - 5 | 999 | 132 | |
NM_001261951.3 | NP_001248880.1 | 4 - 5 | 999 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |