Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutl-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383432.1 | 2 - 3 | 334 | 72 | ||
NM_058397.7 | NP_490798.1 | 7 - 8 | 1606 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |