Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bre-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383737.1 | 5 - 6 | 846 | 74 | ||
NM_058471.2 | NP_490872.1 | 5 - 6 | 851 | 74 | ||
GenBank mRNA | AY130767.1 | - | 4 - 5 | 753 | 74 | |
AY533306.1 | - | 4 - 5 | 753 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |