Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zwl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001262221.3 | NP_001249150.1 | 6 - 7 | 1323 | 86 | |
NM_001262222.4 | NP_001249151.1 | 6 - 7 | 1319 | 86 | ||
NM_001306389.3 | NP_001293318.1 | 2 - 3 | 357 | 86 | ||
NM_001306390.3 | NP_001293319.1 | 2 - 3 | 351 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |