Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dmsr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058591.6 | NP_490992.2 | 1 - 2 | 83 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Caenorhabditis elegans Probe spans exons |