Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cogc-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026635.4 | NP_001021806.1 | 3 - 4 | 425 | 94 | |
NM_001383914.1 | 3 - 4 | 412 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |