Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sep-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058759.6 | NP_491160.1 | 7 - 8 | 2010 | 64 | |
GenBank mRNA | AF399825.1 | - | 6 - 7 | 1987 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |