Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026325.2 | NP_001021496.1 | 22 - 23 | 6032 | 72 | |
NM_001026327.4 | NP_001021498.1 | 6 - 7 | 1566 | 72 | ||
NM_001026328.3 | NP_001021499.1 | 3 - 4 | 1129 | 72 | ||
NM_001026329.6 | NP_001021500.1 | 4 - 5 | 1232 | 72 | ||
NM_001262583.3 | NP_001249512.1 | 4 - 5 | 1225 | 72 | ||
NM_001262584.2 | NP_001249513.1 | 6 - 7 | 1613 | 72 | ||
NM_001375256.3 | NP_001361940.1 | 6 - 7 | 1360 | 72 | ||
NM_001381834.2 | NP_001367474.1 | 22 - 23 | 5920 | 72 | ||
GenBank mRNA | AF048834.1 | - | 22 - 23 | 6032 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |