Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025813.2 | NP_001020984.1 | 7 - 8 | 1249 | 64 | |
NM_001025814.4 | NP_001020985.1 | 7 - 8 | 1058 | 64 | ||
NM_001025817.4 | NP_001020988.1 | 7 - 8 | 1058 | 64 | ||
NM_001025818.3 | NP_001020989.1 | 7 - 8 | 1058 | 64 | ||
NM_001356807.3 | NP_001343717.1 | 6 | 1058 | 64 | ||
NM_001356808.3 | NP_001343716.1 | 6 | 1058 | 64 | ||
NM_001356809.3 | NP_001343714.1 | 6 | 1058 | 64 | ||
NM_001356810.3 | NP_001343718.1 | 6 | 1058 | 64 | ||
NM_001356811.3 | NP_001343715.1 | 6 | 1058 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Both primers and probe map within a single exon |