Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025814.4 | NP_001020985.1 | 38 - 39 | 20993 | 75 | |
NM_001025818.3 | NP_001020989.1 | 36 - 37 | 19073 | 75 | ||
NM_001306524.2 | NP_001293453.1 | 4 - 5 | 1024 | 75 | ||
NM_001306525.3 | NP_001293454.1 | 4 - 5 | 1073 | 75 | ||
NM_001356807.3 | NP_001343717.1 | 37 | 18833 | 75 | ||
NM_001356808.3 | NP_001343716.1 | 38 | 19274 | 75 | ||
NM_001356809.3 | NP_001343714.1 | 37 | 20927 | 75 | ||
NM_001356812.3 | NP_001343617.1 | 29 - 30 | 19574 | 75 | ||
NM_001356813.3 | NP_001343719.1 | 28 - 29 | 17414 | 75 | ||
NM_001356814.3 | NP_001343711.1 | 29 - 30 | 17855 | 75 | ||
NM_001356815.3 | NP_001343712.1 | 28 - 29 | 19508 | 75 | ||
NM_001356816.3 | NP_001343713.1 | 27 - 28 | 17654 | 75 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |