Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nab-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379751.2 | NP_001367535.1 | 3 - 4 | 311 | 52 | |
NM_001379754.2 | NP_001367538.1 | 3 - 4 | 309 | 52 | ||
NM_001382892.1 | 3 - 4 | 308 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |