Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058953.3 | NP_491354.2 | 3 - 4 | 342 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |