Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026429.3 | NP_001021600.1 | 12 - 13 | 2884 | 98 | |
NM_001262699.2 | NP_001249628.1 | 15 - 16 | 2778 | 98 | ||
NM_001372632.2 | NP_001360115.1 | 15 - 16 | 2770 | 98 | ||
NM_001382937.1 | 11 - 12 | 2675 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |