Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026429.3 | NP_001021600.1 | 9 - 10 | 2406 | 149 | |
NM_001262699.2 | NP_001249628.1 | 12 - 13 | 2300 | 149 | ||
NM_001372632.2 | NP_001360115.1 | 12 - 13 | 2292 | 149 | ||
NM_001382937.1 | 8 - 9 | 2197 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |