Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kca-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059042.6 | NP_491443.3 | 2 - 3 | 724 | 130 | |
GenBank mRNA | DQ178243.1 | - | 2 - 3 | 726 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |