Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gnrr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001262791.3 | NP_001249720.1 | 3 - 4 | 662 | 127 | |
NM_001262792.3 | NP_001249721.1 | 7 - 8 | 1156 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |