Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: imb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059076.7 | NP_491477.2 | 8 - 9 | 1794 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |