Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dyf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059093.7 | NP_491494.2 | 11 - 12 | 1547 | 82 | |
NM_182063.7 | NP_871863.1 | 9 - 10 | 1328 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |