Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dyf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059093.7 | NP_491494.2 | 4 - 5 | 550 | 98 | |
NM_182063.7 | NP_871863.1 | 2 - 3 | 331 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |