Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hcp-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059136.4 | NP_491537.1 | 6 - 7 | 2190 | 99 | |
NM_059137.5 | NP_491538.1 | 6 - 7 | 2186 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |