Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: obr-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380388.1 | NP_001367357.1 | 1 - 2 | 286 | 76 | |
NM_059290.7 | NP_491691.1 | 6 - 7 | 1123 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |