Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313481.3 | NP_001300410.1 | 14 | 3677 | 70 | |
NM_001313482.3 | NP_001300411.1 | 14 - 15 | 3776 | 70 | ||
NM_001313483.4 | NP_001300412.1 | 13 - 14 | 3780 | 70 | ||
NM_059306.6 | NP_491707.2 | 13 - 14 | 3617 | 70 | ||
GenBank mRNA | EF205592.1 | - | 13 - 14 | 3605 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |