Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: avr-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383179.1 | 9 - 10 | 937 | 57 | ||
NM_001383181.1 | 7 - 8 | 1100 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |