Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028198.5 | NP_001023369.1 | 5 - 6 | 577 | 66 | |
NM_001307204.3 | NP_001294133.1 | 2 - 3 | 157 | 66 | ||
NM_001307205.2 | NP_001294134.1 | 6 - 7 | 818 | 66 | ||
NM_001307206.3 | NP_001294135.1 | 5 - 6 | 571 | 66 | ||
NM_001307207.3 | NP_001294136.1 | 3 - 4 | 337 | 66 | ||
NM_001380356.1 | NP_001368431.1 | 2 - 3 | 151 | 66 | ||
NM_001383138.1 | 6 - 7 | 905 | 66 | |||
NM_001383139.1 | 2 - 3 | 407 | 66 | |||
NM_171396.7 | NP_741462.1 | 6 - 7 | 804 | 66 | ||
NM_171397.8 | NP_741463.1 | 3 - 4 | 331 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |