Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spp-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171399.6 | NP_741465.1 | 2 - 3 | 414 | 70 | |
NM_171400.5 | NP_741466.1 | 2 - 3 | 408 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |