Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lam-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060374.5 | NP_492775.2 | 8 - 9 | 5991 | 89 | |
GenBank mRNA | AF074902.1 | - | 8 - 9 | 5991 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |