Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exc-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027868.5 | NP_001023039.1 | 13 - 14 | 2182 | 95 | |
NM_001307099.3 | NP_001294028.1 | 12 - 13 | 1981 | 95 | ||
NM_001307100.4 | NP_001294029.1 | 14 - 15 | 2418 | 95 | ||
GenBank mRNA | AB060647.1 | - | 13 - 14 | 2235 | 95 | |
AF339059.1 | - | 13 - 14 | 2235 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |