Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exc-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027868.5 | NP_001023039.1 | 8 - 9 | 1117 | 67 | |
NM_001307099.3 | NP_001294028.1 | 7 - 8 | 916 | 67 | ||
NM_001307100.4 | NP_001294029.1 | 9 - 10 | 1353 | 67 | ||
GenBank mRNA | AB060647.1 | - | 8 - 9 | 1170 | 67 | |
AF339059.1 | - | 8 - 9 | 1170 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |