Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071892.9 | NP_504293.2 | 3 - 4 | 505 | 62 | |
NM_171452.2 | NP_741531.1 | 3 - 4 | 532 | 62 | ||
NM_171927.3 | NP_741532.1 | 3 - 4 | 524 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |