Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mec-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028946.6 | NP_001024117.2 | 27 - 28 | 5230 | 110 | |
NM_001028947.3 | NP_001024118.1 | 24 - 25 | 5054 | 110 | ||
NM_001028949.3 | NP_001024120.1 | 18 - 19 | 4004 | 110 | ||
NM_001373047.2 | NP_001359916.1 | 25 - 26 | 5170 | 110 | ||
NM_001380680.1 | NP_001367617.1 | 26 - 27 | 5210 | 110 | ||
NM_171477.7 | NP_741559.3 | 27 - 28 | 5266 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |