Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spas-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269186.3 | NP_001256115.1 | 1 - 2 | 135 | 63 | |
NM_171501.6 | NP_741586.1 | 1 - 2 | 132 | 63 | ||
NR_136364.1 | 1 - 2 | 121 | 63 | |||
GenBank mRNA | AB287436.1 | - | 1 - 2 | 121 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |