Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136431.4 | NP_001129903.1 | 10 - 11 | 8959 | 97 | |
NM_001269453.3 | NP_001256382.1 | 8 - 9 | 8710 | 97 | ||
NM_001269454.2 | NP_001256383.1 | 10 - 11 | 9201 | 97 | ||
NM_001373124.2 | NP_001360523.1 | 11 | 9268 | 97 | ||
NM_001383415.1 | 10 - 11 | 9207 | 97 | |||
NM_171544.7 | NP_741632.2 | 11 - 12 | 9383 | 97 | ||
GenBank mRNA | AF053496.1 | - | 10 - 11 | 8959 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |