Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136431.4 | NP_001129903.1 | 12 - 13 | 11387 | 69 | |
NM_001269453.3 | NP_001256382.1 | 10 - 11 | 11138 | 69 | ||
NM_001269454.2 | NP_001256383.1 | 12 - 13 | 11629 | 69 | ||
NM_001373124.2 | NP_001360523.1 | 13 | 11696 | 69 | ||
NM_001383415.1 | 12 - 13 | 11635 | 69 | |||
NM_171544.7 | NP_741632.2 | 13 - 14 | 11811 | 69 | ||
GenBank mRNA | AF053496.1 | - | 12 - 13 | 11387 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |