Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hot-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136425.3 | NP_001129897.1 | 4 - 5 | 390 | 110 | |
NM_171552.4 | NP_741640.1 | 4 - 5 | 513 | 110 | ||
GenBank mRNA | AF324053.1 | - | 4 - 5 | 507 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |