Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171588.6 | NP_741678.1 | 1 - 2 | 159 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |