Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aex-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029171.7 | NP_001024342.1 | 13 - 14 | 2759 | 101 | |
NM_171618.5 | NP_741710.1 | 13 - 14 | 2757 | 101 | ||
GenBank mRNA | U93842.1 | - | 13 - 14 | 2779 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |