Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029702.5 | NP_001024873.1 | 2 - 3 | 226 | 84 | |
NM_001374979.2 | NP_001362027.1 | 2 - 3 | 201 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |