Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375019.2 | NP_001361932.1 | 3 | 214 | 70 | |
NM_001375020.2 | NP_001362134.1 | 2 - 3 | 206 | 70 | ||
NM_001375021.2 | NP_001361933.1 | 5 - 6 | 1762 | 70 | ||
NM_001375022.2 | NP_001362133.1 | 5 - 6 | 1758 | 70 | ||
NM_001375024.2 | NP_001362136.1 | 4 - 5 | 1683 | 70 | ||
NM_001383564.1 | 5 - 6 | 1760 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |