Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chtl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171686.7 | NP_741790.2 | 3 - 4 | 174 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |