Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glr-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383601.1 | 4 - 5 | 1462 | 89 | ||
NM_171712.5 | NP_741822.2 | 10 - 11 | 1999 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |