Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171727.4 | NP_741838.1 | 3 - 4 | 596 | 102 | |
GenBank mRNA | AF082011.1 | - | 3 - 4 | 596 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |