Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pkc-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029345.6 | NP_001024516.1 | 9 - 10 | 1495 | 67 | |
NM_001029346.4 | NP_001024517.1 | 9 - 10 | 1496 | 67 | ||
NM_001129650.5 | NP_001123122.1 | 11 - 12 | 1840 | 67 | ||
NM_001373417.3 | NP_001360728.1 | 11 | 1602 | 67 | ||
NM_001373418.2 | NP_001360727.1 | 10 - 11 | 1833 | 67 | ||
NM_001373419.3 | NP_001359883.1 | 10 - 11 | 1680 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |