Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047832.3 | NP_001041297.1 | 2 - 3 | 211 | 71 | |
NM_001381114.2 | NP_001366649.1 | 2 - 3 | 756 | 71 | ||
NM_171782.6 | NP_741908.1 | 1 - 2 | 186 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |