Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047832.3 | NP_001041297.1 | 6 - 7 | 1123 | 75 | |
NM_001381114.2 | NP_001366649.1 | 6 - 7 | 1668 | 75 | ||
NM_001383653.1 | 6 - 7 | 1150 | 75 | |||
NM_171782.6 | NP_741908.1 | 5 - 6 | 1098 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |