Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379816.2 | NP_001367451.1 | 11 - 12 | 2061 | 64 | |
NM_001379817.1 | NP_001367450.1 | 10 - 11 | 2397 | 64 | ||
NM_066340.6 | NP_498741.3 | 12 - 13 | 2294 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |