Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zen-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028138.2 | NP_001023309.1 | 6 - 7 | 2158 | 76 | |
NM_001028139.3 | NP_001023310.1 | 6 - 7 | 2154 | 76 | ||
NM_001028140.5 | NP_001023311.1 | 6 - 7 | 2164 | 76 | ||
NM_001372836.1 | NP_001360082.1 | 6 - 7 | 2164 | 76 | ||
NM_001372837.3 | NP_001360296.1 | 6 - 7 | 2158 | 76 | ||
NM_001372838.2 | NP_001360295.1 | 7 | 2162 | 76 | ||
GenBank mRNA | AF057567.1 | - | 6 - 7 | 2149 | 76 | |
AF057568.1 | - | 6 - 7 | 2149 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |